Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Rabies Virus
- Antigen symbol:Rabies Virus
- Clonality:Monoclonal
- Clone:Rab-50
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Rabies virus of strains SAD-Vnukovo, Pitman Moore
- Form:Liquid
- Antigen synonyms:Rabies Virus
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Purified virions of SAD-Vnukovo strain followed by Pitman-Moore strain.
- Tested applications:ICC
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [Rab-50] antibody to Rabies virus for ELISA, ICC and IF with samples derived from Rabies virus of strains SAD-Vnukovo. Pitman Moore..
- Validated Applications: ELISA, ICC, IF
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml, IF: 1 to 2 µg/ml
Type: Primary
Antigen: Rabies Virus
Clonality: Monoclonal
Clone: Rab-50
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Rabies virus of strains SAD-Vnukovo, Pitman Moore