Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:D6S221E
- Antigen symbol:MHC Class II
- Clonality:Monoclonal
- Clone:MK-D6
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:Liquid
- Gene ID:UniprotID# P01904, P04224
- Antigen synonyms:DPB1|DMA|DPB1_HUMAN|DRB|DPB 1|DP beta 1 chain|DP(W4) beta chain|DP beta 1|DMB
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Spleen cells from B10.D2 mice.
- Tested applications:Functional studies
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [MK-D6] antibody to MHC Class II for Functional Studies, Flow Cytometry and IF with samples derived from mouse.
- Validated Applications: Functional Studies, Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 0,5 to 1 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: MHC Class II
Clonality: Monoclonal
Clone: MK-D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse