Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-Enterovirus 71 3D Rabbit Polyclonal Antibody
Catalog # ANTIA309772-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Enterovirus 71 3D Rabbit Polyclonal Antibody
Catalog # ANTIA309772-100
Supplier:  Antibodies.com
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Enterovirus 71 3D
  • Antigen symbol:
    Enterovirus 71 3D
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Enterovirus 71
  • Western blot:
    Yes
  • Form:
    Liquid
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% thiomersal
  • Molecular weight:
    51 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant protein of Enterovirus EV71 3D.
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to Enterovirus 71 3D for WB with samples derived from Enterovirus 71.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:1,000

Type: Primary
Antigen: Enterovirus 71 3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Enterovirus 71