Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Cytokeratin 17 Rabbit Recombinant Antibody [clone: KRT17/8320R]
Anti-Cytokeratin 17 Rabbit Recombinant Antibody [clone: KRT17/8320R]
  ANTIA314858-100
New Product
Anti-Cytokeratin 17 Rabbit Recombinant Antibody [clone: KRT17/8320R]
  ANTIA314858-100
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    Cytokeratin 17
  • Antigen symbol:
    Cytokeratin 17
  • Clonality:
    Recombinant
  • Clone:
    KRT17/8320R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q04695
  • Antigen synonyms:
    keratin 17 epitope S2|CK-17|Cytokeratin 17|Keratin|39.1|K1C17_HUMAN|keratin 17 epitope S1|Cytokeratin-17|K17|CK 17
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    46 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment of human KRT17 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [KRT17/8320R] antibody to Cytokeratin 17 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 17
Clonality: Recombinant
Clone: KRT17/8320R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human