Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:A disintegrin and metalloproteinase domain 15
- Antigen symbol:ADAM15
- Clonality:Monoclonal
- Clone:ARC1419
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q13444
- Antigen synonyms:ADAM metallopeptidase domain 15|disintegrin-like|and cysteine-rich protein 15|A disintegrin and metalloproteinase domain 15 (metargidin)|ADAM 15|Disintegrin and metalloproteinase domain-containing protein 15|ADA15_HUMAN|EC 3.4.24.|ADAM15
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:100 kDa
- Sequence:RHIRRRRDVVTETKTVELVIVADHSEAQKYRDFQHLLNRTLEVALLLDTFFRPLNVRVALVGLEAWTQRDLVEISPNPAVTLENFLHWRRAHLLPRLPHDS
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 200-300 of human ADAM15 (Q13444)
- Tested applications:IHC, ICC/IF
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC1419] antibody to ADAM15 for WB, IHC and ICC/IF with samples derived from human, mouse and rat.
- Validated applications: WB, IHC, ICC/IF
- Recommended dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: ADAM15
Clonality: Monoclonal
Clone: ARC1419
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat