Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:CD204
- Antigen symbol:Macrophage Scavenger Receptor I
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# P21757
- Antigen synonyms:Macrophage scavenger receptor types I and II|Macrophage scavenger receptor 1|Macrophage acetylated LDL receptor I and II|CD24|Msr 1|Macrophage scavenger receptor type III|CD204 antigen|Macrophage Scavenger Receptor I|MSR1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:49 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from the Internal region of human MSR1 (amino acids 241-290)
- Tested applications:IHC
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to MSR1 for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC-P: 1:50 to 1:300, ELISA: 1:10000
Type: Primary
Antigen: Macrophage Scavenger Receptor I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat