Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Slc6a6 Rabbit Polyclonal Antibody
Anti-Slc6a6 Rabbit Polyclonal Antibody
Catalog # ANTIA96617-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Slc6a6 Rabbit Polyclonal Antibody
Catalog # ANTIA96617-100
Supplier:  ANTIBODIES.COM
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MGC10619
  • Antigen symbol:
    Slc6a6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P31641
  • Antigen synonyms:
    SC6A6_HUMAN|OTTHUMP00000208177|Slc6a6|Sodium- and chloride-dependent taurine transporter|TAUT|MGC131729|Solute carrier family 6 member 6|OTTHUMP00000208180|Solute carrier family 6 (neurotransmitter transporter, taurine), member 6
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    69 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human SLC6A6 (amino acids 561-610)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to SLC6A6 for IHC and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: IHC, ELISA
  • Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000

Type: Primary
Antigen: Slc6a6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat