Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:CD 4
- Antigen symbol:CD4
- Clonality:Monoclonal
- Clone:MEM-16
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P01730
- Antigen synonyms:CD4 antigen (p55)|CD4_HUMAN|CD4 antigen|CD4 (L3T4)|CD4 receptor|CD4mut|CD4 molecule|CD4+ Lymphocyte deficiency, included|CD4
- Storage buffer:Supplied in tris buffered saline, pH 8,0, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:HPB cell line (human peripheral blood leukemia T-cells)
- Purification:Purified by precipitation and chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [MEM-16] antibody to CD4 for flow cytometry and IP with samples derived from human.
- Validated applications: Flow cytometry, IP
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: MEM-16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human