Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Cambridge pathology 1 antigen
- Antigen symbol:CD52
- Clonality:Recombinant
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Recombinant
- Isotype:IgG1
- Reactivity:Human
- Form:Lyophilized
- Gene ID:UniprotID# P31358
- Antigen synonyms:CD 52|CD52 antigen|CAMPATH 1|CD52_HUMAN|CAMPATH-1 antigen|CD52|CDw52|CD52 molecule|CAMPATH 1 antigen
- Storage buffer:Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Storage temperature:Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Concentration:Reconstitution dependent
- Shipping temperature:Shipped on blue ice at 4 °C
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant humanized monoclonal [Alemtuzumab Biosimilar] antibody to CD52 for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: CD52
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human