Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:CADH2_HUMAN
- Antigen symbol:N Cadherin
- Clonality:Monoclonal
- Clone:MNCD2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Chicken,Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P19022
- Antigen synonyms:Cadherin 2 type 1|Cadherin 2|Cadherin 2, type 1, N-cadherin (neuronal)|Cadherin2|Calcium dependent adhesion protein neuronal|CD325|Cadherin 2 N cadherin neuronal|Cadherin-2|Cadherin 2 type 1 N cadherin neuronal
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:99,76 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 308 - 597, of mouse CDH2 protein. The epitope is the extracellular domain of N Cadherin
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rat monoclonal [MNCD2] antibody to N Cadherin for Flow Cytometry, WB and IHC-P with samples derived from Human, Mouse and Chicken.
- Validated applications: Flow cytometry, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: N Cadherin
Clonality: Monoclonal
Clone: MNCD2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human;Mouse;Chicken