About this item
Mouse monoclonal [4D2] antibody to PEA15 for WB, ICC/IF and IHC with samples derived from Human, Rat, Mouse and Bovine.
- Validated applications: WB, ICC/IF, IHC
- Recommended dilutions: WB: 1:1000 to 1:2000, ICC/IF: 1:1000 to 1:2000, IHC: 1:1000 to 1:2000
Type: Primary
Antigen: PEA15
Clonality: Monoclonal
Clone: 4D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mouse, Bovine
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- ANTIA270553-50
- ANTIA270553-100
- Antigen symbol:
- PEA15
- PEA15
- Antigen name:
- 15 kDa phosphoprotein enriched in astrocytes
- 15 kDa phosphoprotein enriched in astrocytes
- Antigen synonyms:
- HUMMAT 1H|Homolog of mouse MAT 1 oncogene|Astrocytic phosphoprotein PEA 15|Astrocytic phosphoprotein PEA15|Homolog of mouse MAT1 oncogene|HMAT 1|HMAT1|HUMMAT1H|Astrocytic phosphoprotein PEA-15
- HUMMAT 1H|Homolog of mouse MAT 1 oncogene|Astrocytic phosphoprotein PEA 15|Astrocytic phosphoprotein PEA15|Homolog of mouse MAT1 oncogene|HMAT 1|HMAT1|HUMMAT1H|Astrocytic phosphoprotein PEA-15
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 4D2
- 4D2
- Reactivity:
- Human, Bovine, Rat, Mouse
- Human, Bovine, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# Q15121
- UniprotID# Q15121
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant full-length human PEA15, expressed in and purified from E. coli.
- Recombinant full-length human PEA15, expressed in and purified from E. coli.
- Form:
- Liquid
- Liquid
- Molecular weight:
- 15 kDa
- 15 kDa
- Purification:
- Immunogen affinity purification
- Immunogen affinity purification
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Supplied in Phosphate Buffered Saline with 50% Glycerol and 5mM Sodium Azide
- Supplied in Phosphate Buffered Saline with 50% Glycerol and 5mM Sodium Azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested applications:
- ICC/IF, IHC
- ICC/IF, IHC