Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:18A2
- Antigen symbol:S100A4
- Clonality:Recombinant
- Clone:S100A4/2750R
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P26447
- Antigen synonyms:Fibroblast specific protein 1 (FSP1)|Fibroblast specific protein|CAPL|calcium Placental protein|Calvasculin|Leukemia multidrug resistance associated protein|FSP1|42A|Fibroblast specific protein 1
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:10 - 12 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 1 - 200, of human S100A4 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [S100A4/2750R] antibody to S100A4 for WB, Flow Cytometry, IF and IHC-P with samples derived from Human and Mouse.
- Validated applications: WB, Flow cytometry, IF, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: S100A4
Clonality: Recombinant
Clone: S100A4/2750R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse