Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-NAPSIN A Rabbit Recombinant Antibody [clone: NAPSA/1865R]
Anti-NAPSIN A Rabbit Recombinant Antibody [clone: NAPSA/1865R]
Catalog # ANTIA250675-100
Supplier:  Antibodies.com
New Product
undefined
Anti-NAPSIN A Rabbit Recombinant Antibody [clone: NAPSA/1865R]
Catalog # ANTIA250675-100
Supplier:  Antibodies.com
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Asp 4
  • Antigen symbol:
    NAPSIN A
  • Clonality:
    Recombinant
  • Clone:
    NAPSA/1865R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O96009
  • Antigen synonyms:
    Kidney derived aspartic protease like protein|NAPA|Kdap|Aspartyl protease 4|KAP|NAP1|Napsa|NAPSA_HUMAN|ASP4
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    37 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 189 - 299, of human Napsin A protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [NAPSA/1865R] antibody to NAPSIN A for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: NAPSIN A
Clonality: Recombinant
Clone: NAPSA/1865R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human