Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Cortisol Binding Globulin Rabbit Polyclonal Antibody
Anti-Cortisol Binding Globulin Rabbit Polyclonal Antibody
Catalog # ANTIA11880-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Cortisol Binding Globulin Rabbit Polyclonal Antibody
Catalog # ANTIA11880-100
Supplier:  Antibodies.com
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CBG
  • Antigen symbol:
    Cortisol Binding Globulin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P08185
  • Antigen synonyms:
    Corticosteroid-binding globulin|_HUMAN|Cortisol Binding Globulin|serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 6|corticosteroid binding globulin|serine (or cysteine) proteinase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 6|Serpin A6|Transcortin|Serpina6
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    45 kDa
  • Sequence:
    SLELLESFSADIKHYYESEVLAMNFQDWATASRQINSYVKNKTQGKIVDLFSGLDSPAILVLVNYIFFKGTWTQPFDLASTREENFYVDETTVVKVPMMLQSSTISYLHDAELPCQLVQMNYVGNGTVFFILPDKGKMNTVIAALSRDTINRWSAGLTSSQVDLYIPKVTISGVYDLGDVLEEMGIADLFTNQANFSRITQDAQLKSSKVVHKAVLQLNEEGVDTAGSTGVTLNLTSKPIILRFNQPFIIMIFDHFTWSSLFLARVMNPV
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 136 - 405 of human SERPINA6 (NP_001747.2)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to Cortisol Binding Globulin for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: Cortisol Binding Globulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human