Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:CD7
- Antigen symbol:CD7
- Clonality:Monoclonal
- Clone:MEM-186
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P09564
- Antigen synonyms:CD7 molecule|CD7_HUMAN|LEU 9|CD7 antigen (p41)|LEU9|p41 protein|CD7 antigen|GP40|T cell antigen CD7
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human acute myelogenous leukaemia cell line KG-1
- Purification:This antibody is conjugated with tandem dye PE-Cyanine 7 under optimum conditions The conjugate is purified by size-exclusion chromatography
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [MEM-186] antibody to CD7 (PE-Cyanine 7) for Flow Cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: MEM-186
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human