Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CXCR4 Mouse Monoclonal Antibody [clone: 12G5] (PE-Cyanine 7)
Anti-CXCR4 Mouse Monoclonal Antibody [clone: 12G5] (PE-Cyanine 7)
  ANTIA121987-100
New Product
undefined
Anti-CXCR4 Mouse Monoclonal Antibody [clone: 12G5] (PE-Cyanine 7)
  ANTIA121987-100
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    C-X-C chemokine receptor type 4
  • Antigen symbol:
    CXCR4
  • Clonality:
    Monoclonal
  • Clone:
    12G5
  • Conjugation:
    PE (Phycoerythrin)-Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P61073
  • Antigen synonyms:
    CXCR4_HUMAN|Chemokine (C X C motif) receptor 4|CXCR-4|CXCR4|CXC-R4|D2S201E|CD184 antigen|Chemokine CXC Motif Receptor 4|CD184
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    CP-MAC infected Sup-T1 cells
  • Purification:
    This antibody is conjugated with tandem dye PE-Cyanine 7 under optimum conditions The conjugate is purified by size-exclusion chromatography
  • Pack type:
    Glass bottle
  • Pk:
    100 T

 

 

Mouse monoclonal [12G5] antibody to CXCR4 (PE-Cyanine 7) for Flow Cytometry with samples derived from human and Non-Human Primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 12G5
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human;Non-Human Primates