Anti-CD83 Mouse Monoclonal Antibody [clone: HB15e] (PE-Cyanine 5)
Catalog # ANTIA122073-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:B cell activation 45kDa cell surface glycoprotein Ig superfamily
- Antigen symbol:CD83
- Clonality:Monoclonal
- Clone:HB15e
- Conjugation:PE (Phycoerythrin)/Cy5®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Non-human primates,Human
- Form:Liquid
- Gene ID:UniprotID# Q01151
- Antigen synonyms:BL11|CD83 antigen|B-cell activation protein|CD83 molecule|CD83|B cell activation protein|CD83_HUMAN|BL11 PEN|CD83 antigen (activated B lymphocytes, immunoglobulin superfamily)
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human CD83 transfected Cos cells
- Purification:This antibody is conjugated with tandem dye PE-Cyanine 5 under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [HB15e] antibody to CD83 (PE-Cyanine 5) for Flow Cytometry with samples derived from human and Non-Human Primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: HB15e
Conjugation: PE-Cyanine 5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-Human Primates