Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-N Cadherin Rabbit Recombinant Antibody [clone: CDH2/8998R]
Anti-N Cadherin Rabbit Recombinant Antibody [clone: CDH2/8998R]
Catalog # ANTIA316746-100
Supplier:  Antibodies.com
New Product
Anti-N Cadherin Rabbit Recombinant Antibody [clone: CDH2/8998R]
Catalog # ANTIA316746-100
Supplier:  Antibodies.com
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CADH2_HUMAN
  • Antigen symbol:
    N Cadherin
  • Clonality:
    Recombinant
  • Clone:
    CDH2/8998R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P19022
  • Antigen synonyms:
    Cadherin 2 type 1|Cadherin 2|Cadherin 2, type 1, N-cadherin (neuronal)|Cadherin2|Calcium dependent adhesion protein neuronal|CD325|Cadherin 2 N cadherin neuronal|Cadherin-2|Cadherin 2 type 1 N cadherin neuronal
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    130 - 140 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 605-905) of human CDH2 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [CDH2/8998R] antibody to N Cadherin for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: N Cadherin
Clonality: Recombinant
Clone: CDH2/8998R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human