Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CD99 Rabbit Recombinant Antibody [clone: MIC2/8119R]
Anti-CD99 Rabbit Recombinant Antibody [clone: MIC2/8119R]
  ANTIA316141-100
New Product
Anti-CD99 Rabbit Recombinant Antibody [clone: MIC2/8119R]
  ANTIA316141-100
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    CD99
  • Antigen symbol:
    CD99
  • Clonality:
    Recombinant
  • Clone:
    MIC2/8119R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P14209
  • Antigen synonyms:
    CD99|CD99_HUMAN|Antigen identified by monoclonal antibodies 12E7, F21 and O13|12E7|CD99 molecule|Cell surface antigen O13|Antigen identified by monoclonal 12E7, Y homolog|Cell surface antigen 12E7|CD99 antigen|Cell surface antigen HBA 71
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    27 - 32 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 1-185) of human MIC2 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [MIC2/8119R] antibody to CD99 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: CD99
Clonality: Recombinant
Clone: MIC2/8119R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human