Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-MGMT Rabbit Monoclonal Antibody [clone: MGMT/8364R]
Anti-MGMT Rabbit Monoclonal Antibody [clone: MGMT/8364R]
Catalog # ANTIA315305-100
Supplier:  Antibodies.com
New Product
Anti-MGMT Rabbit Monoclonal Antibody [clone: MGMT/8364R]
Catalog # ANTIA315305-100
Supplier:  Antibodies.com
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    6 O methylguanine DNA methyltransferase
  • Antigen symbol:
    MGMT
  • Clonality:
    Monoclonal
  • Clone:
    MGMT/8364R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P16455
  • Antigen synonyms:
    AI267024|Methylated DNA protein cysteine methyltransferase|Agat|MGMT|Methylated-DNA--protein-cysteine methyltransferase|MGC107020|6-O-methylguanine-DNA methyltransferase|EC 2.1.1.63|AGT
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
  • Molecular weight:
    21,646 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 1-100) of human MGMT. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit monoclonal [MGMT/8364R] antibody to MGMT for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: MGMT
Clonality: Monoclonal
Clone: MGMT/8364R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human