Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Fc epsilon RI
- Antigen symbol:FCER1A
- Clonality:Monoclonal
- Clone:CRA1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino acids 26 - 110
- Form:Liquid
- Gene ID:UniprotID# P12319
- Antigen synonyms:alpha|Fc fragment of IgE, high affinity I, receptor for, alpha subunit|Fc fragment of IgE, high affinity I, receptor for|Fc IgE receptor, alpha polypeptide|alpha-chain|Fc fragment of IgE high affinity I receptor for alpha polypeptide|Fc IgE receptor alpha polypeptide|Fc IgE receptor, alpha chain|alpha chain
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol (filter-sterilised, without sodium azide and carrier free)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant extracellular portion of human FCER1A protein (corresponding to a.a. Met-26-197, where signal peptide is 1-25).
- Tested applications:IHC-P, IHC-Fr
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:50 µG
Mouse monoclonal (CRA1) antibody to FCER1A (FITC) for WB, FC, IHC-P, IHC-Fr and IF with samples derived from human.
- Validated applications: WB, FC, IHC-P, IHC-Fr, IF
- Recommended dilutions: IF: 2 to 10 µg/ml, FC: 2 to 10 µg/ml
Type: Primary
Antigen: FCER1A
Clonality: Monoclonal
Clone: CRA1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Amino acids 26 to 110
Host: Mouse
Isotype: IgG2b
Reactivity: Human