Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:GluN1
- Antigen symbol:NMDAR1
- Clonality:Monoclonal
- Clone:S308-48
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P35439
- Antigen synonyms:N methyl D aspartate receptor subunit NR1|MRD8|N methyl D aspartate receptor channel subunit zeta 1|Glutamate [NMDA] receptor subunit zeta-1|N methyl D aspartate receptor|glutamate receptor ionotropic, NMDA 1|Glutamate receptor ionotropic, N-methyl-D aspartate, subunit 1|Grin1|Glutamate receptor ionotropic N methyl D aspartate 1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:~105 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein corresponding to the extracellular N - terminus, amino acids 42 - 361, of rat NR1 protein.
- Tested applications:IHC, ICC/IF
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [S308-48] antibody to NMDAR1 for WB, IHC and ICC/IF with samples derived from human, mouse and rat.
- Validated applications: WB, IHC, ICC/IF
- Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100
Type: Primary
Antigen: NMDAR1
Clonality: Monoclonal
Clone: S308-48
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat