About this item
Recombinant rabbit monoclonal [DM205] antibody to ANGPTL3 for ELISA and Flow cytometry with samples derived from human.
- Validated applications: ELISA, Flow cytometry
- Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100
Type: Primary
Antigen: ANGPTL3
Clonality: Recombinant
Clone: DM205
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- ANTIA318504-100
- ANTIA318504-500
- Antigen symbol:
- ANGPTL3
- ANGPTL3
- Antigen name:
- ANG 5
- ANG 5
- Antigen synonyms:
- Angiopoietin 5|Angiopoietin-5|ANG5|Angiopoietin-related protein 3|ANGL3_HUMAN|ANG-5|Angiopoietin like 3|Angiopoietin related protein 3|Angiopoietin-like protein 3
- Angiopoietin 5|Angiopoietin-5|ANG5|Angiopoietin-related protein 3|ANGL3_HUMAN|ANG-5|Angiopoietin like 3|Angiopoietin related protein 3|Angiopoietin-like protein 3
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DM205
- DM205
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# Q9Y5C1
- UniprotID# Q9Y5C1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Form:
- lyophilised
- lyophilised
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
- Storage temperature:
- Shipped at 4 °C. lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C