Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Competitive
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Luteinizing Hormone beta
- Description:Rat luteinizing hormone beta ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Rat Luteinizing Hormone beta ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Luteinizing Hormone beta. No significant cross-reactivity or interference between Luteinizing Hormone beta and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,156 - 10 ng/ml
- Storage temperature:4 °C
- Sample volume:50 μl
- Sensitivity:0,094 ng/ml
- Regulatory status:RUO
Rat Luteinizing Hormone beta ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rat Luteinizing Hormone beta in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 0,156 to 10 ng/ml
- Sensitivity: 0,094 ng/ml
- Sample type: Serum, plasma, tissue homogenates and other biological fluids
- Assay precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat Luteinizing Hormone beta ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of rat Luteinizing Hormone beta in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Luteinizing Hormone beta antigen. During the incubation, Luteinizing Hormone beta present in the samples or standards competes with the fixed amount of immobilised Luteinizing Hormone beta for binding sites on the Biotinylated Anti-Luteinizing Hormone beta Antibody. The more Luteinizing Hormone beta present in a sample or standard, the less Biotinylated Anti-Luteinizing Hormone beta Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Luteinizing Hormone beta Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Luteinizing Hormone beta present in each sample or standard. The concentration of Luteinizing Hormone beta can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.