Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pack type:Vial
- Pk:3
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Storage conditions:Freeze (<−15 °C)
- Protein synonyms:Cationised ovalbumin
- Protein/peptide name:Maleimide Activated cOVA
- Shipping temperature:RT
- Pk Info:3×2 mg
Maleimide Activated cOVA is a sulfhydryl-reactive form specifically designed for efficient hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens.
- Designed for heightened immunogen response
Cationised Ovalbumin (cOVA) is synthesised by treating native ovalbumin with ethylenediamine, which replaces the majority of negatively charged carboxyl groups with positively charged primary amines.