Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Mouse SDF4 ELISA Kit

Mouse SDF4 ELISA Kit

Mouse SDF4 ELISA kit, 96 Tests
Catalog # ANTIA77297-96
Supplier:   Antibodies.com
Mouse SDF4 ELISA Kit
Catalog # ANTIA77297-96
Supplier:  Antibodies.com
CAS Number:  
Mouse SDF4 ELISA Kit
Mouse SDF4 ELISA Kit

Mouse SDF4 ELISA kit, 96 Tests

CAS Number:  
Price
£532.00
Log in for availability
1 * 96 Tests
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Mouse SDF4 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse SDF4 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0,313 to 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse SDF4 employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse SDF4 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for SDF4 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SDF4 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-SDF4 Antibody, which binds the captured SDF4 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of SDF4 captured in each well. The concentration of SDF4 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Mouse SDF4 ELISA kit
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    0,313 - 20 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Mouse SDF4 ELISA kit exhibits high specificity and excellent specificity for the detection of mouse SDF4. No significant cross-reactivity or interference between SDF4 and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    ANTIA77297-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,188 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    SDF4
  • Regulatory status:
    RUO

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.