Order Entry
Northern Ireland
ContactUsLinkComponent
Mouse LTBR ELISA Kit
Mouse LTBR ELISA Kit
  ANTIA312518-96
New Product
 :  
undefined
Mouse LTBR ELISA Kit
  ANTIA312518-96
New Product
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    LTBR
  • Description:
    Mouse LTBR ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    125 - 2000 ng/L
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    9,96 ng/L
  • Regulatory status:
    RUO

 

 

Mouse LTBR ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse LTBR in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 125 to 2000 ng/L
  • Sensitivity: 9,96 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse LTBR ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse LTBR in serum, plasma or other biological fluids. An antibody specific for LTBR has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the LTBR present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-LTBR antibody, which also binds the LTBR present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-LTBR antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-LTBR antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of LTBR captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of LTBR in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.