Order Entry
Northern Ireland
ContactUsLinkComponent
Camel Gastrin ELISA Kit
Camel Gastrin ELISA Kit
Catalog # ANTIA302588-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Camel Gastrin ELISA Kit
Catalog # ANTIA302588-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Other
  • Target protein:
    Gastrin
  • Description:
    Camel Gastrin ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross reactivity:
    Camel Gastrin ELISA kit exhibits high specificity and excellent specificity for the detection of camel Gastrin. No significant cross-reactivity or interference between Gastrin and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,313 - 20 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Sensitivity:
    0,188 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Camel Gastrin ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of camel Gastrin in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection range: 0,313 to 20 pg/ml
  • Sensitivity: 0,188 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Camel Gastrin ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of camel Gastrin in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Gastrin antigen. During the incubation, Gastrin present in the samples or standards competes with the fixed amount of immobilised Gastrin for binding sites on the Biotinylated Anti-Gastrin antibody. The more Gastrin present in a sample or standard, the less Biotinylated Anti-Gastrin antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Gastrin antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Gastrin present in each sample or standard. The concentration of Gastrin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.