Order Entry
Northern Ireland
ContactUsLinkComponent
Mouse ATP1A2 ELISA Kit
Mouse ATP1A2 ELISA Kit
Catalog # ANTIA314327-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Mouse ATP1A2 ELISA Kit
Catalog # ANTIA314327-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    ATP1A2
  • Description:
    Mouse ATP1A2 ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    150 - 2400 ng/L
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    9,94 ng/L
  • Regulatory status:
    RUO

Specifications

About this item

Mouse ATP1A2 ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse ATP1A2 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse ATP1A2 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse ATP1A2 in serum, plasma or other biological fluids. An antibody specific for ATP1A2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ATP1A2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-ATP1A2 antibody, which also binds the ATP1A2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-ATP1A2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-ATP1A2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of ATP1A2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of ATP1A2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.