Human CD31 ELISA Kit
Human CD31 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CD31 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Detection range: 0,781 to 50 ng/ml
- Sensitivity: 0,469 ng/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Human CD31 ELISA kit (A74705) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human CD31 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for CD31 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CD31 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-CD31 Antibody, which binds the captured CD31 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of CD31 captured in each well. The concentration of CD31 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
- Pk:96 Tests
- Description:Human CD31 ELISA kit
- Primary antibody reactivity:Human
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Sample volume:100 μl
- Detection range:0,781 - 50 ng/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Sandwich
- Time to Results:4 h 30 min
- Cross reactivity:Human CD31 ELISA kit exhibits high specificity and excellent specificity for the detection of human CD31. No significant cross-reactivity or interference between CD31 and analogues was observed.
- Assay Principle:Quantitative
- Cat. No.:ANTIA74705-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:0,469 ng/ml
- Format:Pre-coated
- Target protein:CD31
- Regulatory status:RUO
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



