Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:hnRNP A1
- Description:Mouse hnRNP A1 ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Mouse hnRNP A1 ELISA kit exhibits high specificity and excellent specificity for the detection of mouse hnRNP A1. No significant cross-reactivity or interference between hnRNP A1 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:39,063 - 2500 pg/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:23,438 pg/ml
- Regulatory status:RUO
Specifications
About this item
Mouse hnRNP A1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse hnRNP A1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Detection range: 39,063 to 2500 pg/ml
- Sensitivity: 23,438 pg/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Mouse hnRNP A1 ELISA kit (A74818) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse hnRNP A1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for hnRNP A1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the hnRNP A1 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-hnRNP A1 Antibody, which binds the captured hnRNP A1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of hnRNP A1 captured in each well. The concentration of hnRNP A1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.