Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Asph
- Description:Mouse ASPH ELISA kit
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:15 - 240 ng/L
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:0,56 ng/L
- Regulatory status:RUO
Specifications
About this item
Mouse Asph ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Asph in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse Asph ELISA kit (A313577) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Asph in serum, plasma or other biological fluids. An antibody specific for Asph has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Asph present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Asph antibody, which also binds the Asph present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Asph antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Asph antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Asph captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Asph in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.