Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Prothrombin Fragment 1 + 2
- Description:Rat prothrombin fragment 1 + 2 ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Rat Prothrombin Fragment 1 + 2 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Prothrombin Fragment 1 + 2. No significant cross-reactivity or interference between Prothrombin Fragment 1 + 2 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:46,875 - 3000 pg/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:28,125 pg/ml
- Regulatory status:RUO
Rat Prothrombin Fragment 1 + 2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Prothrombin Fragment 1 + 2 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 46,875 to 3000 pg/ml
- Sensitivity: 28,125 pg/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat Prothrombin Fragment 1 + 2 ELISA kit (A79306) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Prothrombin Fragment 1 + 2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Prothrombin Fragment 1 + 2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Prothrombin Fragment 1 + 2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Prothrombin Fragment 1 + 2 Antibody, which binds the captured Prothrombin Fragment 1 + 2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Prothrombin Fragment 1 + 2 captured in each well. The concentration of Prothrombin Fragment 1 + 2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.