Order Entry
Northern Ireland
ContactUsLinkComponent
Mouse HMGB2 ELISA Kit
Mouse HMGB2 ELISA Kit
Catalog # ANTIA310090-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Mouse HMGB2 ELISA Kit
Catalog # ANTIA310090-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    HMGB2
  • Description:
    Mouse HMGB2 ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    1,5 - 24 ng/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    0,056 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse HMGB2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse HMGB2 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 1,5 to 24 ng/mL
  • Sensitivity: 0,056 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Mouse HMGB2 ELISA kit (A310090) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse HMGB2 in serum, plasma or other biological fluids. An antibody specific for HMGB2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the HMGB2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-HMGB2 antibody, which also binds the HMGB2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-HMGB2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-HMGB2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of HMGB2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of HMGB2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.