Order Entry
Northern Ireland
ContactUsLinkComponent
Mouse Cx40/GJA5 ELISA Kit
Mouse Cx40/GJA5 ELISA Kit
Catalog # ANTIA76389-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Mouse Cx40/GJA5 ELISA Kit
Catalog # ANTIA76389-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Cx40/GJA5
  • Description:
    Mouse Cx40/GJA5 ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Mouse Cx40/GJA5 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Cx40/GJA5. No significant cross-reactivity or interference between Cx40/GJA5 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    19,531 - 1250 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    11,719 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse Cx40/GJA5 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Cx40/GJA5 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 19,531 to 1250 pg/ml
  • Sensitivity: 11,719 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse Cx40/GJA5 ELISA Kit (A76389) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Cx40/GJA5 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Cx40/GJA5 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cx40/GJA5 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Cx40/GJA5 Antibody, which binds the captured Cx40/GJA5 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Cx40/GJA5 captured in each well. The concentration of Cx40/GJA5 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.