Order Entry
Northern Ireland
ContactUsLinkComponent
Rabbit Platelet Activating Factor ELISA Kit
Rabbit Platelet Activating Factor ELISA Kit
Catalog # ANTIA75691-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Rabbit Platelet Activating Factor ELISA Kit
Catalog # ANTIA75691-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rabbit
  • Target protein:
    Platelet Activating Factor
  • Description:
    Rabbit platelet activating factor ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Rabbit Platelet Activating Factor ELISA kit exhibits high specificity and excellent specificity for the detection of rabbit Platelet Activating Factor. No significant cross-reactivity or interference between Platelet Activating Factor and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,156 - 10 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Sensitivity:
    0,094 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Rabbit Platelet Activating Factor ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rabbit Platelet Activating Factor in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0,156 to 10 ng/ml
  • Sensitivity: 0,094 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Rabbit Platelet Activating Factor ELISA kit (A75691) employs the competitive enzyme immunoassay technique for the quantitative measurement of rabbit Platelet Activating Factor in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Platelet Activating Factor antigen. During the incubation, Platelet Activating Factor present in the samples or standards competes with the fixed amount of immobilised Platelet Activating Factor for binding sites on the Biotinylated Anti-Platelet Activating Factor Antibody. The more Platelet Activating Factor present in a sample or standard, the less Biotinylated Anti-Platelet Activating Factor Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Platelet Activating Factor Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Platelet Activating Factor present in each sample or standard. The concentration of Platelet Activating Factor can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.