Order Entry
Northern Ireland
ContactUsLinkComponent
Mouse Isoleucyl tRNA Synthetase ELISA Kit
Mouse Isoleucyl tRNA Synthetase ELISA Kit
Catalog # ANTIA78274-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Mouse Isoleucyl tRNA Synthetase ELISA Kit
Catalog # ANTIA78274-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Isoleucyl tRNA Synthetase
  • Description:
    Mouse isoleucyl tRNA synthetase ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Mouse Isoleucyl tRNA Synthetase ELISA kit exhibits high specificity and excellent specificity for the detection of mouse Isoleucyl tRNA Synthetase. No significant cross-reactivity or interference between Isoleucyl tRNA Synthetase and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,625 - 40 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    0,375 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse Isoleucyl tRNA Synthetase ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Isoleucyl tRNA Synthetase in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0,625 to 40 ng/ml
  • Sensitivity: 0,375 ng/ml
  • Sample type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse Isoleucyl tRNA Synthetase ELISA kit (A78274) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Isoleucyl tRNA Synthetase in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Isoleucyl tRNA Synthetase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Isoleucyl tRNA Synthetase present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Isoleucyl tRNA Synthetase Antibody, which binds the captured Isoleucyl tRNA Synthetase present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Isoleucyl tRNA Synthetase captured in each well. The concentration of Isoleucyl tRNA Synthetase can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.