Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Hemoglobin Subunit alpha
- Description:Human hemoglobin subunit alpha ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Human Hemoglobin Subunit alpha ELISA kit exhibits high specificity and excellent specificity for the detection of human Hemoglobin Subunit alpha. No significant cross-reactivity or interference between Hemoglobin Subunit alpha and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:1,563 - 100 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0,938 ng/ml
- Regulatory status:RUO
Specifications
About this item
Human Hemoglobin Subunit alpha ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Hemoglobin Subunit alpha in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 1,563 to 100 ng/ml
- Sensitivity: 0,938 ng/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Hemoglobin Subunit alpha ELISA kit (A78231) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Hemoglobin Subunit alpha in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Hemoglobin Subunit alpha has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Hemoglobin Subunit alpha present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Hemoglobin Subunit alpha Antibody, which binds the captured Hemoglobin Subunit alpha present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Hemoglobin Subunit alpha captured in each well. The concentration of Hemoglobin Subunit alpha can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.