Bottles, Square, HDPE Natural
Supplier: RIXIUS
New Product
Certificates
About this item
Wide-neck square bottles made of HDPE, natural thread.
- Robust HDPE plastic
- Natural plastic (milky-transparent)
- With retracted labeling field
- Tasteless and odorless
- Ideal for filling chemical-technical substances
- With UN approval for solids
- For high-quality powders and salts, filling goods that require UN approval, ideal for retained samples
- Closure: tamper-evident closure blue and red
HDPE is very popular for packaging food, chemicals and cosmetics, among other things. When uncoloured, the plastic appears milky transparent and is hard but flexible at room temperature.
Frequently Bought Together