APC-Cy5.5 Tandem 4 mg/ml, suspension, Sigma-Aldrich®
Catalog # A7472-.5ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:0,5 mL
- Seq:2040
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Colour):Blue
- Appearance (Form) in 60% saturated Ammonium Sulfate and 50 mM Potassium Phosphate, pH 7.0.:Suspension
- Concentration Conforms:4 mg/ml
- Fluorescence, Emission Max:655 - 665 nm
- Relative Quantum Yield:≥1.8
- Wavelength, Lambda Max:645 - 655 nm
- UV Absorbance Ratio, A650/A280:≥4.00
- UV Absorbance Ratio, A650/A615:≥1.30
Specification Test Results
About this item
:
:105000 g/mol
Storage Temperature:Refrigerator
:M-Clarity™ quality level = MQ200