Order Entry
Northern Ireland
ContactUsLinkComponent
Coomassie brilliant blue G-250 concentrate, Sigma-Aldrich®
Coomassie brilliant blue G-250 concentrate, Sigma-Aldrich®
Catalog # B8522-1EA
CAS Number:  
Coomassie brilliant blue G-250 concentrate, Sigma-Aldrich®
Catalog # B8522-1EA
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Suitability for staining protein in polyacrylamide and agarose gels:
    Pass
  • Appearance (Colour):
    Dark Blue
  • Appearance (Form):
    Liquid

Specification Test Results

About this item

Synonyms: Brilliant blue G-250, Serva Blue G, Coomassie Blue G 250, Xylene Brilliant Cyanine G, Brillant blue G, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt, C.I. 42655, Acid Blue 90, Sodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulphonate, Brilliant indocyanine G, Brilliant Blue G
 :
 :854.04 g/mol
UN:3286
 :GHS02
 :GHS06
 :GHS08
CAS Number:6104-58-1
Formula:C₄₇H₄₈N₃NaO₇S₂
MDL Number:MFCD00078482
Flash Pt:9,7 °C (closed cup)
ADR:3 (6.1, 8) II