Order Entry
Northern Ireland
ContactUsLinkComponent
1,9-Dimethyl-Methylene Blue zinc chloride double salt 80% (dye content), Sigma-Aldrich®
Irritant
1,9-Dimethyl-Methylene Blue zinc chloride double salt 80% (dye content), Sigma-Aldrich®
Catalog # 341088-1G
CAS Number:  
Irritant
1,9-Dimethyl-Methylene Blue zinc chloride double salt 80% (dye content), Sigma-Aldrich®
Catalog # 341088-1G
CAS Number:  

Specification Test Results

  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • ICP Major Analysis, Confirms Zinc Component:
    Confirmed
  • Wavelength:
    646 - 652 nm
  • Extinction Coefficient :
    ≥35000
  • Extinction Coefficient:
    ≥18000
  • Extinction Coefficient, c = 0.003 g/L; Water:
    ≥10000
  • Carbon:
    ≥40.5%
  • Nitrogen:
    ≥7.9%
  • Water (by Karl Fischer):
    ≤20.0%

Specification Test Results

About this item