Aroclor 1248 (PCB-mixture, 48% chlorinated)
Catalog # ACCUC-248N
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
Boiling Pt:340 °C (1013 hPa)
Density:1,41 g/cm³ (20 °C)
UN:2315
ADR:9,II
CAS Number:12672-29-6
:1336-36-3