Order Entry
Northern Ireland
ContactUsLinkComponent
Aroclor 1242 (PCB-mixture, 42% chlorinated)
Aroclor 1242 (PCB-mixture, 42% chlorinated)
Catalog # ACCUC-242N
Supplier:  AccuStandard
CAS Number:  
Aroclor 1242 (PCB-mixture, 42% chlorinated)
Catalog # ACCUC-242N
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Boiling Pt:325 °C (1013 hPa)
Melting Pt:–19 °C
Density:1,4 g/cm³ (20 °C)
UN:2315
ADR:9,II
CAS Number:53469-21-9
 :1336-36-3