ContactUsLinkComponent
Thiamine hydrochloride
  MOLE46264200-100G
 :  Molekula
 :  
Thiamine hydrochloride
  MOLE46264200-100G
 :  Molekula
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:
 : Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride
 :
 :337.27 g/mol
 :252
 :1.401 g/cm³ (20 °C)
 :Ambient
 :200-641-8
 :67-03-8
 :C₁₂H₁₈Cl₂N₄OS
 :MFCD00012780
 :13,09366