
Thiamine hydrochloride, (max. 5% H₂O) 99% (dry weight)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-03-8
:
Formula:C₁₂H₁₈Cl₂N₄OS
MDL Number:MFCD00012780
:337.27 g/mol
Merck Index:14,9295
Melting Pt:∼ 250 °C (dec.)
Density:1,401 g/cm³ (20 °C)
EINECS:200-641-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together