Order Entry
Northern Ireland
ContactUsLinkComponent
 
Platinum iridium alloy (90:10 w%) Ø 0,5 mm
  CHMP901134
 :  CHEMPUR
New Product
 :  
Platinum iridium alloy (90:10 w%) Ø 0,5 mm
  CHMP901134
 :  CHEMPUR
New Product
 :  

 

  • Pk:
    10 cm
  • Seq:
    7600

 

 

 :
 :MFCD02091683