Order Entry
Northern Ireland
ContactUsLinkComponent
Carbol fuchsin solution, Sigma-Aldrich®
Irritant
Health
Carbol fuchsin solution, Sigma-Aldrich®
Catalog # HT80116-500ML
CAS Number:  
undefined
Irritant
Health
Carbol fuchsin solution, Sigma-Aldrich®
Catalog # HT80116-500ML
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance (Colour)
    Dark red-purple with metallic sheen:
    Conforms
  • Appearance (Form):
    Solution
  • Appearance (Turbidity):
    Clear
  • Wave Length:
    542 - 552 nm
  • Absorbance:
    0.300 - 0.700
  • Performance:
    Conforms
  • Expiration Date Period
    18 months:
    Complete

Specification Test Results

About this item

 :
 :351.88 g/mol
 :M-Clarity™ quality level = MQ500 (more info.)
 :GHS07
 :GHS08
Storage Temperature:Room Temperature
CAS Number:4197-24-4
Formula:C₂₁H₂₂ClN₃
MDL Number:MFCD00013129