Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-SLFN12L Rabbit Polyclonal Antibody
Anti-SLFN12L Rabbit Polyclonal Antibody
Catalog # PRSI55-507
Supplier:  ProSci Inc.
New Product
undefined
Anti-SLFN12L Rabbit Polyclonal Antibody
Catalog # PRSI55-507
Supplier:  ProSci Inc.
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Schlafen family member 12-like
  • Antigen symbol:
    SLFN12L
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    Amino Acid Range = 95-123
  • Form:
    Liquid
  • Gene ID:
    Q6IEE8
  • Antigen synonyms:
    SLFN12L
  • Modification:
    None
  • Storage buffer:
    Supplied in PBS with 0,09% (W/V) sodium azide
  • Molecular weight:
    67 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This SLFN12L antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 95-123 amino acids from the N-terminal region of human SLFN12L.
  • Purification:
    This antibody is purified through a protein A column, followed by peptide affinity purification.
  • Size:
    400 µl
  • Pk:
    400 µl

Specifications

About this item

For WB starting dilution is: 1:1000 For FACS starting dilution is: 1:10~50

Type: Primary
Antigen: SLFN12L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human