Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:adhesion G protein-coupled receptor E1
- Antigen symbol:Adgre1
- Clonality:Recombinant
- Clone:Cl:A3-1 (recombinant version)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Cross adsorption:No
- Form:Liquid
- Gene ID:13733
- Antigen synonyms:f-480|EMR1 receptor|mouse macrophage marker|EGF-like module receptor 1|Em Gpf4|f 480|f480|f4-80|Ly-71|Cell surface glycoprotein F4/80|f4 80|TM7|f-4/80|EGF-like module
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Thioglycollate stimulated peritoneal macrophages of mouse origin.
- Size:0,2 mg
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: adhesion G protein-coupled receptor E1
Clonality: Recombinant
Clone: Cl:A3-1 (recombinant version)
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Mouse